Learn More
Biolegend Alexa Fluor 647 anti-human CD137 (4-1BB)
Supplier:  Biolegend 309823
Encompass_Preferred
Alexa Fluor 647 anti-human CD137 (4-1BB) [4B4-1]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Chimpanzee, Baboon, Cynomolgus, Rhesus; Apps: FC; Size: 25 tests
Catalog No. 50-207-3422