Learn More
Biolegend Alexa Fluor 647 anti-human CD147
Supplier:  Biolegend 306210
Encompass_Preferred
Alexa Fluor 647 anti-human CD147 [HIM6]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Chimpanzee, Baboon, Capuchin Monkey; Apps: FC, ICC, IHC-P; Size: 100 tests
Catalog No. 50-166-332