Learn More
Biolegend Alexa Fluor 647 anti-human CD163
Supplier:  Biolegend 326508
Encompass_Preferred
Alexa Fluor 647 anti-human CD163 [RM3/1]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. 50-167-332