Learn More
Biolegend Alexa Fluor 647 anti-human CD19
Supplier:  Biolegend 302222
Encompass_Preferred
Alexa Fluor 647 anti-human CD19 [HIB19]; Isotype: Mouse IgG1, κ; Reactivity: Human, Chimpanzee, Rhesus; Apps: FC, IHC-F; Size: 25 tests
Catalog No. 50-165-837