Learn More
Biolegend Alexa Fluor 647 anti-human CD197 (CCR7)
Supplier:  Biolegend 353217
Encompass_Preferred
Alexa Fluor 647 anti-human CD197 (CCR7) [G043H7]; Isotype: Mouse IgG2a, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: FC; Size: 25 tests
Catalog No. 50-168-770