Learn More
Biolegend Alexa Fluor 647 anti-human CD200 (OX2)
Supplier:  Biolegend 329214
Encompass_Preferred
Alexa Fluor 647 anti-human CD200 (OX2) [OX-104]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon; Apps: FC; Size: 100 tests
Catalog No. 50-207-5845