Learn More
Biolegend Alexa Fluor 647 anti-human CD268 (BAFF-R)
Supplier:  Biolegend 316914
Encompass_Preferred
Alexa Fluor 647 anti-human CD268 (BAFF-R) [11C1]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: FC; Size: 100 tests
Catalog No. 50-166-894