Learn More
Biolegend Alexa Fluor 647 anti-human CD3
Supplier:  Biolegend 300422
Encompass_Preferred
Alexa Fluor 647 anti-human CD3 [UCHT1]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Chimpanzee; Apps: FC, ICC, IHC-F; Size: 25 tests
Catalog No. 50-165-570