Learn More
Biolegend Alexa Fluor 647 anti-human CD303 (BDCA-2)
Supplier:  Biolegend 354217
Encompass_Preferred
Alexa Fluor 647 anti-human CD303 (BDCA-2) [201A]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: FC; Size: 25 tests
Catalog No. 50-207-7330