Learn More
Biolegend Alexa Fluor 647 anti-human CD31
Supplier:  Biolegend 303112
Encompass_Preferred
Alexa Fluor 647 anti-human CD31 [WM59]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: FC; Size: 100 tests
Catalog No. 50-165-962