Learn More
Biolegend Alexa Fluor 647 anti-human CD319 (CRACC)
Supplier:  Biolegend 331819
Encompass_Preferred
Alexa Fluor 647 anti-human CD319 (CRACC) [162.1]; Isotype: Mouse IgG2b, κ; Reactivity: Human, Cynomolgus; Apps: FC; Size: 25 tests
Catalog No. 50-207-6059