Learn More
Biolegend Alexa Fluor 647 anti-human CD5
Supplier:  Biolegend 300616
Encompass_Preferred
Alexa Fluor 647 anti-human CD5 [UCHT2]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Chimpanzee, Capuchin Monkey, Common Marmoset, Cynomolgus, Rhesus; Apps: FC; Size: 100 tests
Catalog No. 50-165-612