Learn More
Biolegend Alexa Fluor 647 anti-human CD56 (NCAM)
Supplier:  Biolegend 304612
Encompass_Preferred
Alexa Fluor 647 anti-human CD56 (NCAM) [MEM-188]; Isotype: Mouse IgG2a, κ; Reactivity: Human, Cross-Reactivity: Cattle (Bovine, Cow); Apps: FC; Size: 100 tests
Catalog No. 50-166-142