Learn More
Biolegend Alexa Fluor 647 anti-human CD66b
Supplier:  Biolegend 305109
Encompass_Preferred
Alexa Fluor 647 anti-human CD66b [G10F5]; Isotype: Mouse IgM, κ; Reactivity: Human, Cross-Reactivity: Chimpanzee; Apps: FC, IHC-P; Size: 25 tests
Catalog No. 50-207-3187