Learn More
Biolegend Alexa Fluor 647 anti-human CD85j (ILT2)
Supplier:  Biolegend 333709
Encompass_Preferred
Alexa Fluor 647 anti-human CD85j (ILT2) [GHI/75]; Isotype: Mouse IgG2b, κ; Reactivity: Human; Apps: FC; Size: 25 tests
Catalog No. 50-167-737