Learn More
Biolegend Alexa Fluor 647 anti-human IL-1β
Supplier:  Biolegend 508207
Encompass_Preferred
Alexa Fluor 647 anti-human IL-1β [JK1B-1]; Isotype: Mouse IgG; Reactivity: Human; Apps: ICFC; Size: 25 tests
Catalog No. 50-170-050