Learn More
Biolegend Alexa Fluor 647 anti-human IL-21
Supplier:  Biolegend 513006
Encompass_Preferred
Alexa Fluor 647 anti-human IL-21 [3A3-N2]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Rhesus; Apps: ICFC; Size: 100 tests
Catalog No. 50-170-153