Learn More
Biolegend Alexa Fluor 647 anti-human Perforin
Supplier:  Biolegend 308109
Encompass_Preferred
Alexa Fluor 647 anti-human Perforin [dG9]; Isotype: Mouse IgG2b, κ; Reactivity: Human, Cross-Reactivity: Bovine (Cow, Cattle); Apps: ICFC; Size: 25 tests
Catalog No. 50-166-458