Learn More
Biolegend Alexa Fluor 647 anti-Human Podoplanin
Supplier:  Biolegend 337007
Encompass_Preferred
Alexa Fluor 647 anti-Human Podoplanin [NC-08]; Isotype: Rat IgG2a, λ; Reactivity: Human; Apps: FC; Size: 25 tests
Catalog No. 50-167-886