Learn More
Biolegend Alexa Fluor 647 anti-human TCR γ/δ
Supplier:  Biolegend 331214
Encompass_Preferred
Alexa Fluor 647 anti-human TCR γ/δ [B1]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Pigtailed Macaque, Rhesus; Apps: FC; Size: 100 tests
Catalog No. 50-167-632