Learn More
Biolegend Alexa Fluor 647 anti-human Cadherin 11
Supplier:  Biolegend 368713
Encompass_Preferred
Alexa Fluor 647 anti-human Cadherin 11 [16G5]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: IHC-P; Size: 25 ug
Catalog No. 50-207-8458