Learn More
Biolegend Alexa Fluor 647 anti-human CD11b
Supplier:  Biolegend 393110
Encompass_Preferred
Alexa Fluor 647 anti-human CD11b [LM2]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. 50-207-8966