Learn More
Biolegend Alexa Fluor 647 anti-human CD44
Supplier:  Biolegend 397511
Encompass_Preferred
Alexa Fluor 647 anti-human CD44 [C44Mab-5]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC, IHC-P; Size: 25 tests
Catalog No. 50-207-9323