Learn More
Biolegend Alexa Fluor 647 anti-human CD44 isoform 9 (CD44v9)
Supplier:  Biolegend 394407
Encompass_Preferred
Alexa Fluor 647 anti-human CD44 isoform 9 (CD44v9) [RV3]; Isotype: Rat IgG2a, κ; Reactivity: Human; Apps: FC, IHC-P; Size: 25 tests
Catalog No. 50-207-9109