Learn More
Biolegend Alexa Fluor 647 anti-human Cytokeratin 7
Supplier:  Biolegend 601608
Encompass_Preferred
Alexa Fluor 647 anti-human Cytokeratin 7 [W16155A]; Isotype: Rat IgG2a, κ; Reactivity: Human; Apps: ICC; Size: 100 ug
Catalog No. 50-208-6629