Learn More
Biolegend Alexa Fluor 647 anti-human ROR1
Supplier:  Biolegend 357821
Encompass_Preferred
Alexa Fluor 647 anti-human ROR1 [2A2]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 25 tests
Catalog No. 50-207-7720