Learn More
Biolegend Alexa Fluor 647 anti-mouse CD16.2 (FcγRIV)
Supplier:  Biolegend 149526
Encompass_Preferred
Alexa Fluor 647 anti-mouse CD16.2 (FcγRIV) [9E9]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. 50-207-1878