Learn More
Biolegend Alexa Fluor 647 anti-mouse CD285 (TLR5)
Supplier:  Biolegend 148104
Encompass_Preferred
Alexa Fluor 647 anti-mouse CD285 (TLR5) [ACT5]; Isotype: Mouse IgG1, κ; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. 50-207-1779