Learn More
Biolegend Alexa Fluor 647 anti-mouse CD301a (MGL1)
Supplier:  Biolegend 145604
Encompass_Preferred
Alexa Fluor 647 anti-mouse CD301a (MGL1) [LOM-8.7]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. 50-207-1652