Learn More
Biolegend Alexa Fluor 647 anti-mouse/human A2B5
Supplier:  Biolegend 150704
Encompass_Preferred
Alexa Fluor 647 anti-mouse/human A2B5 [105]; Isotype: Mouse IgM, κ; Reactivity: Mouse, Human; Apps: FC; Size: 100 ug
Catalog No. 50-207-1974