Learn More
Biolegend Alexa Fluor 647 anti-mouse Ki-67
Supplier:  Biolegend 652407
Encompass_Preferred
Alexa Fluor 647 anti-mouse Ki-67 [16A8]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: ICFC; Size: 25 ug
Catalog No. 50-605-863