Learn More
Biolegend Alexa Fluor 647 anti-mouse Ly-6C
Supplier:  Biolegend 128009
Encompass_Preferred
Alexa Fluor 647 anti-mouse Ly-6C [HK1.4]; Isotype: Rat IgG2c, κ; Reactivity: Mouse; Apps: FC; Size: 25 ug
Catalog No. 50-164-436