Learn More
Biolegend Alexa Fluor 647 anti-mouse CD11a
Supplier:  Biolegend 101114
Encompass_Preferred
Alexa Fluor 647 anti-mouse CD11a [M17/4]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. 50-162-518