Learn More
Biolegend Alexa Fluor 647 anti-mouse CD144 (VE-cadherin)
Supplier:  Biolegend 138107
Encompass_Preferred
Alexa Fluor 647 anti-mouse CD144 (VE-cadherin) [VECD1]; Isotype: Rat IgG1, κ; Reactivity: Mouse; Apps: FC; Size: 25 ug
Catalog No. 50-165-022