Learn More
Biolegend Alexa Fluor 647 anti-mouse CD155 (PVR)
Supplier:  Biolegend 131529
Encompass_Preferred
Alexa Fluor 647 anti-mouse CD155 (PVR) [TX56]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. 50-207-0945