Learn More
Biolegend Alexa Fluor 647 anti-mouse CD19
Supplier:  Biolegend 115522
Encompass_Preferred
Alexa Fluor 647 anti-mouse CD19 [6D5]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC, IHC-F; Size: 100 ug
Catalog No. 50-207-0193