Learn More
Biolegend Alexa Fluor 647 anti-mouse CD199 (CCR9)
Supplier:  Biolegend 128707
Encompass_Preferred
Alexa Fluor 647 anti-mouse CD199 (CCR9) [CW-1.2]; Isotype: Mouse IgG2a; Reactivity: Mouse; Apps: FC; Size: 25 ug
Catalog No. 50-164-493