Learn More
Biolegend Alexa Fluor 647 anti-mouse CD1d (CD1.1, Ly-38)
Supplier:  Biolegend 123511
Encompass_Preferred
Alexa Fluor 647 anti-mouse CD1d (CD1.1, Ly-38) [1B1]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 25 ug
Catalog No. 50-164-123