Learn More
Biolegend Alexa Fluor 647 anti-mouse CD22
Supplier:  Biolegend 126108
Encompass_Preferred
Alexa Fluor 647 anti-mouse CD22 [OX-97]; Isotype: Rat IgG1, κ; Reactivity: Mouse; Apps: FC, IHC-F; Size: 100 ug
Catalog No. 50-164-279