Learn More
Biolegend Alexa Fluor 647 anti-mouse CD23
Supplier:  Biolegend 101611
Encompass_Preferred
Alexa Fluor 647 anti-mouse CD23 [B3B4]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC, IHC-F; Size: 25 ug
Catalog No. 50-162-579