Learn More
Biolegend Alexa Fluor 647 anti-mouse CD24
Supplier:  Biolegend 101818
Encompass_Preferred
Alexa Fluor 647 anti-mouse CD24 [M1/69]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. 50-162-598