Learn More
Biolegend Alexa Fluor 647 anti-mouse CD25
Supplier:  Biolegend 102020
Encompass_Preferred
Alexa Fluor 647 anti-mouse CD25 [PC61]; Isotype: Rat IgG1, λ; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. 50-162-627