Learn More
Biolegend Alexa Fluor 647 anti-mouse CD31
Supplier:  Biolegend 102415
Encompass_Preferred
Alexa Fluor 647 anti-mouse CD31 [390]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC, IHC-F; Size: 25 ug
Catalog No. 50-162-687