Learn More
Biolegend Alexa Fluor 647 anti-mouse CD326 (Ep-CAM)
Supplier:  Biolegend 118212
Encompass_Preferred
Alexa Fluor 647 anti-mouse CD326 (Ep-CAM) [G8.8]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC, IHC-F; Size: 100 ug
Catalog No. 50-163-761