Learn More
Biolegend Alexa Fluor 647 anti-mouse CD36
Supplier:  Biolegend 102610
Encompass_Preferred
Alexa Fluor 647 anti-mouse CD36 [HM36]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. 50-162-715