Learn More
Biolegend Alexa Fluor 647 anti-mouse CD40
Supplier:  Biolegend 102911
Encompass_Preferred
Alexa Fluor 647 anti-mouse CD40 [HM40-3]; Isotype: Armenian Hamster IgM; Reactivity: Mouse; Apps: FC; Size: 25 ug
Catalog No. 50-206-9691