Learn More
Biolegend Alexa Fluor 647 anti-mouse CD45
Supplier:  Biolegend 103123
Encompass_Preferred
Alexa Fluor 647 anti-mouse CD45 [30-F11]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC, IF, IHC; Size: 25 ug
Catalog No. 50-604-824