Learn More
Biolegend Alexa Fluor 647 anti-mouse CD45RB
Supplier:  Biolegend 103312
Encompass_Preferred
Alexa Fluor 647 anti-mouse CD45RB [C363-16A]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. 50-162-824