Learn More
Biolegend Alexa Fluor 647 anti-mouse CD49e
Supplier:  Biolegend 103811
Encompass_Preferred
Alexa Fluor 647 anti-mouse CD49e [5H10-27(MFR5)]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 25 ug
Catalog No. 50-162-886