Learn More
Biolegend Alexa Fluor 647 anti-mouse CD5
Supplier:  Biolegend 100613
Encompass_Preferred
Alexa Fluor 647 anti-mouse CD5 [53-7.3]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 25 ug
Catalog No. 50-162-470